
Terfenadine

Synonym(s):
α-[4-(1,1-Dimethylethyl)phenyl]-4-(hydroxydiphenylmethyl)-1-piperidinebutanol
Empirical Formula (Hill Notation):
C32H41NO2
CAS Number:
Molecular Weight:
471.67
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3