
Rink amide (aminomethyl)polystyrene

Synonym(s):
4-[(2,4-Dimethoxyphenyl)(Fmoc-amino)methyl]phenoxyacetomido methyl resin, Fmoc Rink amide, Knorr Linker amide resin
Products
Brand
Physical Form
Functional Group
Application
Cleavage Condition
Linker
Chain Type
Reaction Type
Showing 1-2 of 2