
Prasugrel Hydrochloride

Synonym(s):
2-[2-(Acetyloxy)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]-1-cyclopropyl-2-(2-fluorophenyl)ethanone hydrochloride
Empirical Formula (Hill Notation):
C20H20FNO3S · HCl
CAS Number:
Molecular Weight:
409.90
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3