
Pelargonidin 3,5-diglucoside chloride

Synonym(s):
Pelargonin chloride, 3,5-Bis(glucosyloxy)-4′,7-dihydroxyflavylium chloride, Monardin, Pelargonidin 3,5-di-O-glucoside chloride
Empirical Formula (Hill Notation):
C27H31ClO15
CAS Number:
Molecular Weight:
630.98
Products
Formula Weight
Manufacturer
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing