Skip to Content
Merck
PalGly ≥98% (HPLC), solid

PalGly

PalGly ≥98% (HPLC), solid
Synonym(s):
N-(1-Oxohexadecyl)glycine, N-Hexadecanoylglycine, Hexadecanoylaminoacetic acid, N-palmitoyl glycine
Empirical Formula (Hill Notation):
C18H35NO3
CAS Number:
Molecular Weight:
313.48

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing