
Isocarbamide

Empirical Formula (Hill Notation):
C8H15N3O2
CAS Number:
Molecular Weight:
185.22
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing