
Glyceryl trinonadecanoate

Synonym(s):
Trinonadecanoin
Empirical Formula (Hill Notation):
C60H116O6
CAS Number:
Molecular Weight:
933.56
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing