
Glucagon-Like Peptide I human

Synonym(s):
GLP-1, Preproglucagon 72-108
Empirical Formula (Hill Notation):
C186H275N51O59
CAS Number:
Molecular Weight:
4169.48
Products
Product Category
Brand
Formula Weight
Gene Alias
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing