
D-Glucose-13C6

Synonym(s):
D-Glucose-ul-13C, Dextrose-13C6
Empirical Formula (Hill Notation):
13C6H12O6
CAS Number:
Molecular Weight:
186.11
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Special Grade
Technique
Showing 1-3 of 3