Skip to Content
Merck
Cystine NIST® SRM® 143d

Cystine

Cystine NIST® SRM® 143d
Synonym(s):
L-Cystine, (R,R)-3,3′-Dithiobis(2-aminopropionic acid)
Linear Formula:
[-SCH2CH(NH2)CO2H]2
CAS Number:
Molecular Weight:
240.30

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing