Skip to Content
Merck
Cyromazine pharmaceutical secondary standard, certified reference material

Cyromazine

Cyromazine pharmaceutical secondary standard, certified reference material
Synonym(s):
N-Cyclopropyl-1,3,5-triazin-2,4,6-triamine, Cyromazin
Empirical Formula (Hill Notation):
C6H10N6
CAS Number:
Molecular Weight:
166.18

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade