
Clavulanate Lithium

Synonym(s):
Lithium clavulanate
Empirical Formula (Hill Notation):
C8H8LiNO5
CAS Number:
Molecular Weight:
205.09
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing