
Cefazolin Impurity H

Synonym(s):
7-Aminocephalosporanic acid, 7-ACA
Empirical Formula (Hill Notation):
C10H12N2O5S
CAS Number:
Molecular Weight:
272.28
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing