
3-sn-Phosphatidylethanolamine from bovine brain

Synonym(s):
1,2-Diacyl-sn-glycero-3-phosphoethanolamine, L-α-Cephalin, L-α-Phosphatidylethanolamine from bovine brain
CAS Number:
EC Number:
Products
Product Category
Brand
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing