Key Documents
Z743568
PetriStickers
12-sector pie format
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
packaging
pack of 36 ea
manufacturer/tradename
Diversified Biotech PSTK-2020
General description
Quick Reference - Petristickers correspond to wells on a microplate for fast colony identification
High Contrast - Colored grid allows colonies to stand out from the Petri dish background
Updated Design - Rectangular shape leaves ample space above and below grid for writing on the dish
Two Layouts - Reference wells numerically (1-96) or alpha numerically (A1-H12)
Ideally Sized - PetriStickers are 2.8 in. x 1.75 in. (71 mm x 44.5 mm), to fit most common Petri dishes
Legal Information
p3xFLAG is a trademark of Sigma-Aldrich Co. LLC