Key Documents
EP4987000010
Eppendorf® Multipette® E3/E3x/M4
E3 single-channel, with charging cable and Combitips advanced assortment pack (1 Combitip of each size), 1 ul - 50 ml, AC/DC input 100 V / 240 V, 50-60 Hz (EU)
Synonym(s):
Adjustable volume pipette
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
CE compliant
E3 single-channel
with charging cable and Combitips advanced assortment pack (1 Combitip of each size)
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 4987000010 (EU)
measuring range
1 ul - 50 ml
volume
1 μL × 50 mL
battery power
rechargeable
General description
The Multipette E3/E3x in combination with the Combitips® advanced creates a versatile dispensing system that functions according to the positive displacement principle and is therefore capable of pipetting any liquid while maintaining the highest standards for accurate results. The motor driven dispenser gives you full volume control, improved accuracy and reproducibility while reducing physical strain during use. The repetitive dispensing modes contribute to reducing fatigue by decreasing the number of movements for the user; filling the tip once allows dispensing up to 100 times. The system is an essential lab tool ideal for all liquid handling applications, accommodating more than 5,000 different dispensing volumes between 1 μL to 50 mL.
Legal Information
Combitips is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Multipette is a registered trademark of Eppendorf AG