Skip to Content
Merck
All Photos(1)

Documents

681636-M

Millipore

W-13, Hydrochloride

A cell-permeable and reversible calmodulin antagonist that inhibits myosin light chain kinase (IC₅₀ = 58 µM) and Ca2+-calmodulin-dependent phosphodiesterase (IC₅₀ = 68 µM).

Synonym(s):

W-13, Hydrochloride, N-(4-Aminobutyl)-5-chloro-2-naphthalenesulfonamide, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17ClN2O2S · xHCl
CAS Number:
Molecular Weight:
312.81 (free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 35 mg/mL

storage temp.

2-8°C

InChI

1S/C14H17ClN2O2S.ClH/c15-14-5-3-4-11-10-12(6-7-13(11)14)20(18,19)17-9-2-1-8-16;/h3-7,10,17H,1-2,8-9,16H2;1H

InChI key

QKAALLVQBOLELJ-UHFFFAOYSA-N

General description

A cell-permeable and reversible calmodulin antagonist that inhibits Ca2+-calmodulin-dependent phosphodiesterase (IC50 = 68 µM) and myosin light chain kinase (IC50 = 58 µM).
A cell-permeable and reversible calmodulin antagonist that inhibits myosin light chain kinase (IC50 = 58 µM) and Ca2+-calmodulin-dependent phosphodiesterase (IC50 = 68 µM).

Biochem/physiol Actions

Target IC50: 58 µM, 68 µM, against myosin light chain kinase and Ca2+-calmodulin-dependent phosphodiesterase

Warning

Toxicity: Standard Handling (A)

Analysis Note

single spot by TLC

Other Notes

Hidaka, H., and Tanaka, T. 1983. Methods Enzymol. 102, 185.
Tanaka, T., et al. 1983. Pharmacol.26, 249.
Tanaka, T., and Hidaka, H. 1980. J. Biol. Chem. 255, 11078.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service