Merck
  • LIM kinase-2 induces programmed necrotic neuronal death via dysfunction of DRP1-mediated mitochondrial fission.
LIM kinase-2 induces programmed necrotic neuronal death via dysfunction of DRP1-mediated mitochondrial fission.
Cell death and differentiation (2014-02-25)
J-E Kim, H J Ryu, M J Kim, T-C Kang
ABSTRACT
Although the aberrant activation of cell cycle proteins has a critical role in neuronal death, effectors or mediators of cyclin D1/cyclin-dependent kinase 4 (CDK4)-mediated death signal are still unknown. Here, we describe a previously unsuspected role of LIM kinase 2 (LIMK2) in programmed necrotic neuronal death. Downregulation of p27(Kip1) expression by Rho kinase (ROCK) activation induced cyclin D1/CDK4 expression levels in neurons vulnerable to status epilepticus (SE). Cyclin D1/CDK4 complex subsequently increased LIMK2 expression independent of caspase-3 and receptor interacting protein kinase 1 activity. In turn, upregulated LIMK2 impaired dynamic-related protein-1 (DRP1)-mediated mitochondrial fission without alterations in cofilin phosphorylation/expression and finally resulted in necrotic neuronal death. Inhibition of LIMK2 expression and rescue of DRP1 function attenuated this programmed necrotic neuronal death induced by SE. Therefore, we suggest that the ROCK-p27(Kip1)-cyclin D1/CDK4-LIMK2-DRP1-mediated programmed necrosis may be new therapeutic targets for neuronal death.
Sigma-Aldrich
Anti-Cofilin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-LIMK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution