Merck
  • Study of nitro-polycyclic aromatic hydrocarbons in particulate matter in Dongguan.
Study of nitro-polycyclic aromatic hydrocarbons in particulate matter in Dongguan.
Environmental science and pollution research international (2014-03-01)
Shan-Hong Lan, Hui-Xia Lan, Dan Yang, Xiu-Wen Wu
ABSTRACT
Total suspended and size-segregated atmospheric particles were collected in four seasons at three representative points in different functional areas of Dongguan City. The detailed size distributions of six nitro-PAHs [2-nitrofluorene (2-NF), 9-nitroanthracene, 2-nitrofluoranthene (2-NFL), 3-nitrofluoranthene, 1-nitropyrene, and 2-nitropyrene (2-NP)] were determined by high-performance liquid chromatography (HPLC) with UV detection using a binary elution gradient (methanol and water). We used a toxicity assessment based on potency equivalency factors (PEFs) to estimate the inhalation risk of the particulate matter. The results showed that, aside from 2-NF and 2-NFL, the content of the other four nitro-PAHs in the microparticles (<0.4 μm) were more than 20%, a percentage significantly higher than other fractions of particulate matter. The seasonal distribution of nitro-PAHs shows that their concentrations were higher in the winter, while the PAH concentrations were higher in the summer. The study found that secondary formation (2-NFL and 2-NP) had a positive correlation with NO x and NO2, but a negative correlation with O3. The benzo[a]pyrene equivalent (BaPeq) toxicity of particulate matter in Dongguan City ranged from 0.04 to 2.63 ng m(-3), and the carcinogenic index ranged from 0.04 × 10(-6) to 2.39 × 10(-6). These values do not represent a serious threat to human health.
Sigma-Aldrich
3-Nitrofluoranthene, technical grade, 90%
Pricing and availability is not currently available.