Merck
  • Alterations in diurnal rhythmicity in patients treated for nonfunctioning pituitary macroadenoma: a controlled study and literature review.
Alterations in diurnal rhythmicity in patients treated for nonfunctioning pituitary macroadenoma: a controlled study and literature review.
European journal of endocrinology (2014-05-16)
S D Joustra, R D Thijs, R van den Berg, M van Dijk, A M Pereira, G J Lammers, E J W van Someren, J A Romijn, N R Biermasz
ABSTRACT
Patients treated for nonfunctioning pituitary macroadenomas (NFMAs) have fatigue and alterations in sleep characteristics and sleep-wake rhythmicity frequently. As NFMAs often compress the optic chiasm, these complaints might be related to dysfunction of the adjacent suprachiasmatic nucleus (SCN). We aimed to explore whether indirect indices of SCN functioning are altered in the long term after surgery for NFMAs. We studied 17 NFMA patients in long-term remission after transsphenoidal surgery, receiving adequate and stable hormone replacement for hypopituitarism, and 17 control subjects matched for age, gender, and BMI. Indirect indices of SCN function were assessed from 24-h ambulatory recordings of skin and core body temperatures, blood pressure, and salivary melatonin levels. Altered melatonin secretion was defined as an absence of evening rise, considerable irregularity, or daytime values >3 pg/ml. We additionally studied eight patients treated for craniopharyngioma. Distal-proximal skin temperature gradient did not differ between NFMAs and control subjects, but proximal skin temperature was decreased during daytime (P=0.006). Core body temperature and non-dipping of blood pressure did not differ, whereas melatonin secretion was often altered in NFMAs (OR 5.3, 95% CI 0.9-30.6). One or more abnormal parameters (≥2.0 SDS of control subjects) were observed during nighttime in 12 NFMA patients and during daytime in seven NFMA patients. Similar patterns were observed in craniopharyngioma patients. Heterogeneous patterns of altered diurnal rhythmicity in skin temperature and melatonin secretion parameters were observed in the majority of patients treated for NFMAs. On a group level, both NFMA and craniopharyngioma patients showed a lower daytime proximal skin temperature than control subjects, but other group averages were not significantly different. The observations suggest altered function of central (or peripheral) clock machinery, possibly by disturbed entrainment or damage of the hypothalamic SCN by the suprasellar macroadenoma or its treatment.
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
USP
Melatonin, United States Pharmacopeia (USP) Reference Standard