Merck
  • Structure of beta-crystallite assemblies formed by Alzheimer beta-amyloid protein analogues: analysis by x-ray diffraction.
Structure of beta-crystallite assemblies formed by Alzheimer beta-amyloid protein analogues: analysis by x-ray diffraction.
Biophysical journal (1993-02-01)
H Inouye, P E Fraser, D A Kirschner
ABSTRACT
To elucidate the relation between amyloid fibril formation in Alzheimer disease and the primary structure of the beta/A4 protein, which is the major component of the amyloid, we have been investigating the ability of peptides sharing sequences with beta/A4 to form fibrils in vitro. In previous studies we focused on the macroscopic morphology of the assemblies formed by synthetic peptides corresponding in sequence to different regions of this protein. In the present study we analyze the x-ray diffraction patterns obtained from these assemblies. All specimens showed wide angle reflections that could be indexed by an orthogonal lattice of beta-crystallites having unit cell dimensions a = 9.4 A, b = 7 A, and c = 10 A, where a refers to hydrogen bonding direction, b to polypeptide chain direction, and c to intersheet direction. Given the amino acid sequence of beta/A4 as NH2-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIAT-COOH, we found that, based on their orientation and assembly, the analogues could be classified into three groups: Group A, residues 19-28, 13-28, 12-28, 11-28, 9-28, 1-28, 1-38, 1-40, 6-25, 11-25 and 34-42; Group B, residues 18-28, 17-28, and 15-28; and Group C, residues 22-35 and 26-33. For Groups A and C, the sharpest reflections were (h00), indicating that the assemblies were fibrillar, i.e., elongated in a single direction. Lateral alignment of the crystallites in Group A account for its cross-beta pattern, in which the hydrogen bonding (H-bonding) direction is the fiber (rotation) axis. By comparison, the beta-crystallites of Group C had no preferential orientation, thus giving circular scattering. For Group B, the sharpest reflections were (h0l) on the meridian, indicating that the assemblies were plate-like, i.e., extended in two directions. A series of equatorial Bragg reflections having a 40 A period indicated regular stacking of the plates, and the rotation axis was normal to the surface of the plates. Of the Group A peptides, the analogues 11-28 and 6-25 showed intensity maxima on the equator as well as on higher layer lines, indicating that the beta-crystallites are highly ordered relative to one another in the axial, H-bonding direction. This sampling of the layer lines by a larger period (60 A) suggests that the beta-crystallites are arrayed either in cylindrical or small restricted crystalline lattices. Consistent with its electron microscopic images, we modeled the structure as a tube with five or six f,-crystallites constituting the wall and with the individual crystallite, which either rotates freely or is restricted, made of five or fewer beta-pleated sheets. For the Group B peptides, the electron density projection along the b-axis was calculated from the observed intensities using phase combinations from fl-keratin.Amino acid side-chain positions were apparent and, when refined as 4-A-diameter spheres, led to a substantial decrease in the R-factors.For peptide 18-28 the electron density peaks, which are thought to correspond to side chains, were centered 3.3 A from the peptide backbone, whereas for peptides 17-28 and 15-28, these peaks were centered 1 A or more further from the backbone. Peaks having high electron density faced peaks having lower density, suggesting a favorable stereochemical arrangement of the residues. Thus, our analysis of the fiber x-ray patterns from beta/A4 peptides shows the organization of the beta-crystallites that form the wall of the amyloid fibrils as well as possible side-chain interactions.
Sigma-Aldrich
Amyloid β-Protein Fragment 17-28, ≥90% (HPLC), lyophilized powder
Pricing and availability is not currently available.