Merck
  • Behavior of glioblastoma brain tumor stem cells following a suborbital rocket flight: reaching the "edge" of outer space.
Behavior of glioblastoma brain tumor stem cells following a suborbital rocket flight: reaching the "edge" of outer space.
NPJ microgravity (2023-12-19)
Cesar A Garcia, Paola Suárez-Meade, Mieu Brooks, Adip G Bhargav, Michelle L Freeman, Lawrence M Harvey, John Quinn, Alfredo Quiñones-Hinojosa
ABSTRACT
The emerging arena of space exploration has created opportunities to study cancer cell biology in the environments of microgravity and hypergravity. Studying cellular behavior in altered gravity conditions has allowed researchers to make observations of cell function that would otherwise remain unnoticed. The patient-derived QNS108 brain tumor initiating cell line (BTIC), isolated from glioblastoma (GBM) tissue, was launched on a suborbital, parabolic rocket flight conducted by EXOS Aerospace Systems & Technologies. All biologicals and appropriate ground controls were secured post-launch and transported back to our research facility. Cells from the rocket-flight and ground-based controls were isolated from the culture containers and expanded on adherent flasks for two weeks. In vitro migration, proliferation, and stemness assays were performed. Following cell expansion, male nude mice were intracranially injected with either ground-control (GC) or rocket-flight (RF) exposed cells to assess tumorigenic capacity (n = 5 per group). Patient-derived QNS108 BTICs exposed to RF displayed more aggressive tumor growth than the GC cells in vitro and in vivo. RF cells showed significantly higher migration (p < 0.0000) and stemness profiles (p < 0.01) when compared to GC cells. Further, RF cells, when implanted in vivo in the brain of rodents had larger tumor-associated cystic growth areas (p = 0.00029) and decreased survival (p = 0.0172) as compared to those animals that had GC cells implanted.
Sigma-Aldrich
Anti-Human Nuclei Antibody, clone 235-1, Biotin Conjugate, clone 235-1, from mouse, biotin conjugate
Pricing and availability is not currently available.