Merck
Fish Scales Produce Cortisol upon Stimulation with ACTH.
Animals : an open access journal from MDPI (2022-12-24)
Athanasios Samaras, Michail Pavlidis
ABSTRACT
Cortisol concentration in fish scales is a novel and reliable indicator of chronic stress. However, until now cortisol in scales has been considered to be accumulated through the circulation and it has not yet been studied whether it can be de novo produced from cells found in the scales. In the current study, scales of European sea bass, Dicentrarchus labrax, were stimulated in-vitro with a range of concentrations of adrenocorticotropic hormone (ACTH) to investigate if they can produce and release cortisol. Moreover, scales were exposed to a combination of ACTH and metyrapone, an inhibitor of cortisol production, to examine whether cortisol was actually produced in the scales. Results from ACTH administration showed that scales increased their cortisol release in a dose-dependent manner. This effect was reversed when scales were co-incubated with ACTH and metyrapone, indicating that cortisol was produced de novo and not released only upon stimulation with ACTH.
Sigma-Aldrich
Adrenocorticotropic Hormone human, ≥97% (HPLC)