Merck
  • Development and optimisation of a defined high cell density yeast medium.
Development and optimisation of a defined high cell density yeast medium.
Yeast (Chichester, England) (2020-02-18)
Tania Michelle Roberts, Hans-Michael Kaltenbach, Fabian Rudolf
ABSTRACT
Saccharomyces cerevisiae cells grown in a small volume of chemically defined media neither reach the desired cell density nor grow at a fast enough rate to scale down the volume and increase the sample number of classical biochemical assays, as the detection limit of the readout often requires a high number of cells as an input. To ameliorate this problem, we developed and optimised a new high cell density (HCD) medium for S. cerevisiae. Starting from a widely used synthetic medium composition, we systematically varied the concentrations of all components without the addition of other compounds. We used response surface methodology to develop and optimise the five components of the medium: glucose, yeast nitrogen base, amino acids, monosodium glutamate, and inositol. We monitored growth, cell number, and cell size to ensure that the optimisation was towards a greater density of cells rather than just towards an increase in biomass (i.e., larger cells). Cells grown in the final medium, HCD, exhibit growth more similar to the complex medium yeast extract peptone dextrose (YPD) than to the synthetic defined (SD) medium. Whereas the final cell density of HCD prior to the diauxic shift is increased compared with YPD and SD about threefold and tenfold, respectively. We found normal cell-cycle behaviour throughout the growth phases by monitoring DNA content and protein expression using fluorescent reporters. We also ensured that HCD media could be used with a variety of strains and that they allow selection for all common yeast auxotrophic markers.
Sigma-Aldrich
Amylase Activity Assay Kit, sufficient for 100 colorimetric tests