Merck
  • Effects of abomasal infusion of essential fatty acids together with conjugated linoleic acid in late and early lactation on performance, milk and body composition, and plasma metabolites in dairy cows.
Effects of abomasal infusion of essential fatty acids together with conjugated linoleic acid in late and early lactation on performance, milk and body composition, and plasma metabolites in dairy cows.
Journal of dairy science (2020-06-02)
L Vogel, M Gnott, C Kröger-Koch, D Dannenberger, A Tuchscherer, A Tröscher, H Kienberger, M Rychlik, A Starke, L Bachmann, H M Hammon
ABSTRACT
Rations including high amounts of corn silage are currently very common in dairy production. Diets with corn silage as forage source result in a low supply of essential fatty acids, such as α-linolenic acid, and may lead to low conjugated linoleic acid (CLA) production. The present study investigated the effects of abomasal infusion of essential fatty acids, especially α-linolenic acid, and CLA in dairy cows fed a corn silage-based diet on performance, milk composition, including fatty acid (FA) pattern, and lipid metabolism from late to early lactation. Rumen-cannulated Holstein cows (n = 40) were studied from wk 9 antepartum to wk 9 postpartum and dried off 6 wk before calving. The cows were assigned to 1 of 4 treatment groups. Cows were abomasally supplemented with coconut oil (CTRL, 76 g/d), linseed and safflower oil (EFA, 78 and 4 g/d; linseed/safflower oil ratio = 19.5:1; n-6/n-3 FA ratio = 1:3), Lutalin (CLA, 38 g/d; BASF SE, Ludwigshafen, Germany; isomers cis-9,trans-11 and trans-10,cis-12 each 10 g/d) or EFA+CLA. Milk composition was analyzed weekly, and blood samples were taken several times before and after parturition to determine plasma concentrations of metabolites related to lipid metabolism. Liver samples were obtained by biopsy on d 63 and 21 antepartum and on d 1, 28, and 63 postpartum to measure triglyceride concentration. Body composition was determined after slaughter. Supplementation of CLA reduced milk fat concentration, increased body fat mass, and improved energy balance (EB) in late and early lactation, but EB was lowest during late lactation in the EFA group. Cows with CLA treatment alone showed an elevated milk citrate concentration in early lactation, whereas EFA+CLA did not reveal higher milk citrate but did have increased acetone. Milk protein was increased in late lactation but was decreased in wk 1 postpartum in CLA and EFA+CLA. Milk urea was reduced by CLA treatment during the whole period. After calving, the increase of nonesterified fatty acids in plasma was less in CLA groups; liver triglycerides were raised lowest at d 28 in CLA groups. Our data confirm an improved metabolic status with CLA but not with exclusive EFA supplementation during early lactation. Increased milk citrate concentration in CLA cows points to reduced de novo FA synthesis in the mammary gland, but milk citrate was less affected in EFA+CLA cows, indicating that EFA supplementation may influence changes in mammary gland FA metabolism achieved by CLA.
Sigma-Aldrich
Triglyceride Quantification Colorimetric/Fluorometric Kit, sufficient for 100 colorimetric or fluorometric tests
Pricing and availability is not currently available.