Merck
  • Chinese Propolis Prevents Obesity and Metabolism Syndromes Induced by a High Fat Diet and Accompanied by an Altered Gut Microbiota Structure in Mice.
Chinese Propolis Prevents Obesity and Metabolism Syndromes Induced by a High Fat Diet and Accompanied by an Altered Gut Microbiota Structure in Mice.
Nutrients (2020-04-03)
Yufei Zheng, Yuqi Wu, Lingchen Tao, Xi Chen, Trevor Joseph Jones, Kai Wang, Fuliang Hu
ABSTRACT
The increasing incidence of obesity poses a great threat to public health worldwide. Recent reports also indicate the relevance of obesity in metabolic diseases. Chinese propolis (CP), as a well-studied natural nutraceutical, has shown a beneficial effect on alleviating diabetes mellitus. However, few studies have investigated the effect of CP on weight management and energy balance. We examined the beneficial effects of dietary CP on weight in high-fat diet-fed female and male mice and determined whether CP alters gut microbiota. In this study, dietary CP supplementation reduces body weight and improves insulin resistance in high-fat diet (HFD)-fed mice in a dose-dependent manner. CP treatment also reverses liver weight loss and triglyceride accumulation in association with hepatic steatosis. The 16S rRNA analysis of gut microbiota demonstrated that CP treatment modulates the composition in HFD-fed mice. Our study also suggests that male mice were more sensitive to CP treatment than female mice. Taken together, CP supplementation reduces weight gain and reverses gut microbiome dysbiosis induced by HFD. Further, the effects of CP treatment on metabolic biomarkers and microbiome structure differ by gender.
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), Mw 10,000