Merck
  • Effect of anti-angiogenesis induced by chemotherapeutic monotherapy, chemotherapeutic/bisphosphonate combination therapy and anti-VEGFA mAb therapy on tooth extraction socket healing in mice.
Effect of anti-angiogenesis induced by chemotherapeutic monotherapy, chemotherapeutic/bisphosphonate combination therapy and anti-VEGFA mAb therapy on tooth extraction socket healing in mice.
Journal of bone and mineral metabolism (2017-10-19)
Yuri Akita, Shinichiro Kuroshima, Kazunori Nakajima, Hiroki Hayano, Riho Kanai, Muneteru Sasaki, Takashi Sawase
ABSTRACT
Osteonecrosis of the jaw (ONJ), which is a rare but severe adverse effect, mainly occurs in oncology patients receiving chemotherapeutic agents and bisphosphonates. However, the combined impact of chemotherapy and bisphosphonates on wound healing after tooth extraction remains unknown. The aim of this study was to determine the precise etiology of ONJ induced by chemotherapy and bisphosphonate combination therapy. Mice received zoledronate (ZA) monotherapy, cyclophosphamide (CY) monotherapy or CY/ZA combination therapy. The maxillary first molars were extracted 3 weeks after the initiation of drug treatment. Moreover, antivascular endothelial growth factor A (VEGFA) monoclonal antibody (mAb) was administered once every 2 days just after tooth extraction for 2 weeks. Soft and hard tissue wound healing was evaluated 2 and 4 weeks post-extraction using histomorphometry, microcomputed tomography and immunohistochemistry. ZA monotherapy did not induce impaired oral wound healing and ONJ-like lesions 2 and 4 weeks post-extraction, respectively. Tooth extraction socket healing worsened with severe anti-angiogenesis by CY monotherapy and CY/ZA combination therapy 2 weeks post-extraction. However, CY monotherapy rarely induced ONJ-like lesions with severe angiogenesis suppression, whereas CY/ZA combination therapy frequently induced ONJ-like lesions with severe angiogenesis inhibition 4 weeks post-extraction. Interestingly, anti-VEGFA mAb therapy delayed osseous wound healing with normal soft tissue wound healing of tooth extraction sockets, although this therapy significantly suppressed blood vessel formation. Our findings suggest that anti-angiogenesis alone is not the main cause of ONJ-like lesions induced by CY/ZA combination therapy. The combination of suppressed osteoclasts and anti-angiogenesis, in addition to other risk factors such as chemotherapy, may contribute to the development of ONJ.
Sigma-Aldrich
Leukocyte Acid Phosphatase (TRAP) Kit, Select reagents packaged in gelatin capsules
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)