Merck
G-quadruplex-induced instability during leading-strand replication.
The EMBO journal (2011-08-30)
Judith Lopes, Aurèle Piazza, Rodrigo Bermejo, Barry Kriegsman, Arianna Colosio, Marie-Paule Teulade-Fichou, Marco Foiani, Alain Nicolas
ABSTRACT
G-quadruplexes are four-stranded nucleic acid structures whose biological functions remain poorly understood. In the yeast S. cerevisiae, we report that G-quadruplexes form and, if not properly processed, pose a specific challenge to replication. We show that the G-quadruplex-prone CEB1 tandem array is tolerated when inserted near ARS305 replication origin in wild-type cells but is very frequently destabilized upon treatment with the potent Phen-DC(3) G-quadruplex ligand, or in the absence of the G-quadruplex-unwinding Pif1 helicase, only when the G-rich strand is the template of leading-strand replication. The orientation-dependent instability is associated with the formation of Rad51-Rad52-dependent X-shaped intermediates during replication detected by two-dimensional (2D) gels, and relies on the presence of intact G-quadruplex motifs in CEB1 and on the activity of ARS305. The asymmetrical behaviour of G-quadruplex prone sequences during replication has implications for their evolutionary dynamics within genomes, including the maintenance of G-rich telomeres.
Sigma-Aldrich
PhenDC3, ≥97% (HPLC)