![Vitamin E dry powder, (50 g all-rac-a-tocopheryl acetate in 100 g subst), EMPROVE® ESSENTIAL, Ph. Eur., BP, USP](/deepweb/assets/sigmaaldrich/product/structures/200/059/08c94b30-ea4f-4708-9d29-cf7df2e43182/640/08c94b30-ea4f-4708-9d29-cf7df2e43182.png)
Vitamin E
![Vitamin E dry powder, (50 g all-rac-a-tocopheryl acetate in 100 g subst), EMPROVE® ESSENTIAL, Ph. Eur., BP, USP](/deepweb/assets/sigmaaldrich/product/structures/200/059/08c94b30-ea4f-4708-9d29-cf7df2e43182/640/08c94b30-ea4f-4708-9d29-cf7df2e43182.png)
Synonym(s):
(±)-α-Tocopherol, DL-all-rac-α-Tocopherol, Vitamin E
Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Markush Class
Markush Group
Showing 1-3 of 3
Compare
Product Number
Description
Pricing