
Risedronate sodium 2.5-hydrate

Synonym(s):
Risedronic acid monosodium salt hemi(pentahydrate)
Empirical Formula (Hill Notation):
C7H10NNaO7P2 · 2.5H2O
CAS Number:
Molecular Weight:
350.13
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing