
Rifaximin for system suitability

Synonym(s):
Rifaximin, 4-Deoxy-4′-methylpyrido[1′,2′-1,2]imidazo[5,4-c]rifamycin SV, Rifacol
Empirical Formula (Hill Notation):
C43H51N3O11
CAS Number:
Molecular Weight:
785.88
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing