![Lysine acetate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/365/096/44dbc337-66c4-4832-baf6-d8645a660e77/640/44dbc337-66c4-4832-baf6-d8645a660e77.png)
Lysine acetate
![Lysine acetate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/365/096/44dbc337-66c4-4832-baf6-d8645a660e77/640/44dbc337-66c4-4832-baf6-d8645a660e77.png)
Synonym(s):
L-Lysine acetate salt, (S)-2,6-Diaminohexanoic acid
Empirical Formula (Hill Notation):
C6H14N2O2 · C2H4O2
CAS Number:
Molecular Weight:
206.24
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing