![Lovastatin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/292/064/28db6eb4-fc27-4e28-b249-5aff010a5458/640/28db6eb4-fc27-4e28-b249-5aff010a5458.png)
Lovastatin
![Lovastatin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/292/064/28db6eb4-fc27-4e28-b249-5aff010a5458/640/28db6eb4-fc27-4e28-b249-5aff010a5458.png)
Synonym(s):
Lovastatin, Mevinolin
Empirical Formula (Hill Notation):
C24H36O5
CAS Number:
Molecular Weight:
404.54
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing