![Di-[4-(N,N-di-p-tolyl-amino)-phenyl]cyclohexane ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/111/787/16bde1ce-c76d-46d6-9e1f-9ce09f82d038/640/16bde1ce-c76d-46d6-9e1f-9ce09f82d038.png)
Di-[4-(N,N-di-p-tolyl-amino)-phenyl]cyclohexane
![Di-[4-(N,N-di-p-tolyl-amino)-phenyl]cyclohexane ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/111/787/16bde1ce-c76d-46d6-9e1f-9ce09f82d038/640/16bde1ce-c76d-46d6-9e1f-9ce09f82d038.png)
Synonym(s):
4,4′-Cyclohexylidenebis[N,N-bis(4-methylphenyl)benzenamine], TAPC
Empirical Formula (Hill Notation):
C46H46N2
CAS Number:
Molecular Weight:
626.87
Products
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing