
Atropine impurity B

Synonym(s):
Noratropine, N-Demethylatropine
Empirical Formula (Hill Notation):
C16H21NO3
CAS Number:
Molecular Weight:
275.34
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing