Skip to Content
Merck
4-O-Caffeoylquinic acid ≥98.0%

4-O-Caffeoylquinic acid

4-O-Caffeoylquinic acid ≥98.0%
Synonym(s):
4-O-(3,4-Dihydroxycinnamoyl)-D-quinic acid, Cryptochlorogenic acid
Empirical Formula (Hill Notation):
C16H18O9
CAS Number:
Molecular Weight:
354.31

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing