4-Methylumbelliferyl oleate
Synonym(s):
Oleic acid 4-methylumbelliferyl ester
Empirical Formula (Hill Notation):
C28H40O4
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing