
2,4,5-Trichloro-6-hydroxyisophthalonitrile

Synonym(s):
2,4,5-Trichloro-6-hydroxy-1,3-benzenedicarbonitrile, Hydroxychlorothalonil
Empirical Formula (Hill Notation):
C8HCl3N2O
CAS Number:
Molecular Weight:
247.47
Products
Product Category
Brand
Formula Weight
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing