Skip to Content
Merck
All Photos(1)

Key Documents

860560P

Avanti

18:1-12:0 Biotin PS

Avanti Research - A Croda Brand 860560P, powder

Synonym(s):

1-oleoyl-2-(12-biotinyl(aminododecanoyl))-sn-glycero-3-phospho-L-serine (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H86N5O12PS
CAS Number:
Molecular Weight:
964.24
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860560P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860560P

lipid type

omega FAs

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCCCCCCNC(CCCC[C@]1([H])SC[C@@]2([H])[C@]1([H])NC(N2)=O)=O)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

InChI

1S/C46H83N4O12PS.H3N/c1-2-3-4-5-6-7-8-9-10-11-12-14-17-20-23-30-42(52)59-33-37(34-60-63(57,58)61-35-38(47)45(54)55)62-43(53)31-24-21-18-15-13-16-19-22-27-32-48-41(51)29-26-25-28-40-44-39(36-64-40)49-46(56)50-44;/h9-10,37-40,44H,2-8,11-36,47H2,1H3,(H,48,51)(H,54,55)(H,57,58)(H2,49,50,56);1H3/b10-9-;/t37-,38+,39+,40+,44+;/m1./s1

InChI key

FUCJSUOHIUUCPS-FNICJJOFSA-N

General description

18:1-12:0 Biotin PS is a biotinylated lipid.

Application

18:1-12:0 Biotin PS may be used in lipid blot analysis to study Niemann-Pick C2 (NPC2)-lipid interactions.

Packaging

5 mL Clear Glass Sealed Ampule (860560P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular cholesterol trafficking is dependent upon NPC2 interaction with lysobisphosphatidic acid
McCauliff LA, et al.
eLife, 8 (2019)
Peter A Leventis et al.
Annual review of biophysics, 39, 407-427 (2010-03-03)
Phosphatidylserine (PS) is the most abundant negatively charged phospholipid in eukaryotic membranes. PS directs the binding of proteins that bear C2 or gamma-carboxyglutamic domains and contributes to the electrostatic association of polycationic ligands with cellular membranes. Rather than being evenly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service