Merck
  • Genome-wide transcriptional profiling analysis reveals annexin A6 as a novel EZH2 target gene involving gastric cellular proliferation.
Genome-wide transcriptional profiling analysis reveals annexin A6 as a novel EZH2 target gene involving gastric cellular proliferation.
Molecular bioSystems (2015-05-08)
Ying Qi, Xiaoli Zhang, Yani Kang, Jun Wu, Jian Chen, Hua Li, Yan Guo, Bingya Liu, Zhifeng Shao, Xiaodong Zhao
ABSTRACT
A histone methyltransferase enhancer of zeste homologue 2 (EZH2) catalyzes trimethylation at histone H3 lysine27 (H3K27me3) and is frequently dysregulated in a wide range of human cancers. EZH2-mediated gene silencing contributes to carcinogenesis and regulates stem cell maintenance and differentiation; however, the underlining mechanisms remain to be completely understood. Here, we found that downregulation of EZH2 by RNA interference (RNAi) in gastric cancer cells suppresses cell growth, migration, invasion, and induces cell cycle arrest. Transcriptome analysis identified 1223 EZH2 responsive genes upon EZH2 knockdown. These genes are involved in the biological processes of cell cycle, proliferation and metastasis. Particularly, we found that annexin A6 (ANXA6) is a new target of EZH2 and is repressed in gastric cancer cells. Restoration of ANXA6 expression inhibits gastric cellular proliferation. We further demonstrated that EZH2-mediated H3K27me3, rather than promoter DNA methylation, is primarily responsible for ANXA6 inhibition. Taken together, our results provide a framework for understanding EZH2 biology and reveal ANXA6 as a new EZH2 target involving gastric cellular proliferation.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
MISSION® esiRNA, targeting human EZH2
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ezh2