Merck
  • Protective effects of myricitrin against osteoporosis via reducing reactive oxygen species and bone-resorbing cytokines.
Protective effects of myricitrin against osteoporosis via reducing reactive oxygen species and bone-resorbing cytokines.
Toxicology and applied pharmacology (2014-08-19)
Qiang Huang, Bo Gao, Long Wang, Ya-Qian Hu, Wei-Guang Lu, Liu Yang, Zhuo-Jing Luo, Jian Liu
ABSTRACT
Oxidative stress is a crucial pathogenic factor in the development of osteoporosis. Myricitrin, isolated from Myrica cerifera, is a potent antioxidant. We hypothesized that myricitrin possessed protective effects against osteoporosis by partially reducing reactive oxygen species (ROS) and bone-resorbing cytokines in osteoblastic MC3T3-E1 cells and human bone marrow stromal cells (hBMSCs). We investigated myricitrin on osteogenic differentiation under oxidative stress. Hydrogen peroxide (H2O2) was used to establish an oxidative cell injury model. Our results revealed that myricitrin significantly improved some osteogenic markers in these cells. Myricitrin decreased lipid production and reduced peroxisome proliferator-activated receptor gamma-2 (PPARγ2) expression in hBMSCs. Moreover, myricitrin reduced the expression of receptor activator of nuclear factor kappa-B ligand (RANKL) and IL-6 and partially suppressed ROS production. In vivo, we established a murine ovariectomized (OVX) osteoporosis model. Our results demonstrated that myricitrin supplementation reduced serum malondialdehyde (MDA) activity and increased reduced glutathione (GSH) activity. Importantly, it ameliorated the micro-architecture of trabecular bones in the 4th lumbar vertebrae (L4) and distal femur. Taken together, these results indicated that the protective effects of myricitrin against osteoporosis are linked to a reduction in ROS and bone-resorbing cytokines, suggesting that myricitrin may be useful in bone metabolism diseases, particularly osteoporosis.
Sigma-Aldrich
Mouse TRANCE ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Myricitrin, ≥99.0% (HPLC)
Supelco
Myricitrin, analytical standard