Merck
  • Monomeric, one- and two-dimensional networks incorporating (2,6-Me2C6H3S)2Pb building blocks.
Monomeric, one- and two-dimensional networks incorporating (2,6-Me2C6H3S)2Pb building blocks.
Dalton transactions (Cambridge, England : 2003) (2004-10-29)
Sarah E Appleton, Glen G Briand, Andreas Decken, Anita S Smith
ABSTRACT
The amine coordination of lead(II) has been examined through the preparation and structural analysis of Lewis base adducts of bis(thiolato)lead(II) complexes. Reaction of Pb(OAc)(2) with 2,6-dimethylbenzenethiol affords (2,6-Me(2)C(6)H(3)S)(2)Pb (6) in high yield. The solubility of 6 in organic solvents allows for the preparation of the 1:2 Lewis acid-base adduct [(2,6-Me(2)C(6)H(3)S)(2)Pb(py)(2)](7), and 1:1 adducts [(2,6-Me(2)C(6)H(3)S)(2)Pb(micro(2)-bipy)](infinity](8) and [(2,6-Me(2)C(6)H(3)S)(2)Pb(micro(2)-pyr)](infinity)(9)(where py = pyridine, bipy = 4,4'-bipyridyl and pyr = pyrazine) from reaction with an excess of the appropriate amine. In contrast to 7, reaction of (C(6)H(5)S)(2)Pb (1) with pyridine afforded the 2:1 adduct [(C(6)H(5)S)(4)Pb(2)(py)](infinity)(10). Compounds were characterized via elemental analysis, FT-IR, solution (1)H and (13)C[(1)H](6) NMR spectroscopy, and X-ray crystallography (7-10). The structures of 7-9 show the thiolate groups occupying two equatorial positions and two amine nitrogen atoms occupying axial coordination sites, yielding distorted see-saw coordination geometries, or distorted trigonal bipyramids if an equatorial lone pair on lead is considered. The absence of intermolecular contacts in 7 and 8 result in monomeric and one-dimensional polymeric structures, respectively. Weak Pb...S intermolecular contacts in 9 result in the formation of a two-dimensional macrostructure. In contrast, the structure of , shows extensive intermolecular Pb...S interactions, resulting in five- and six-coordinate bonding environments for lead(II), and a complex polymeric structure in the solid state. This demonstrates the ability of the 2,6-dimethylphenylthiolate ligand to limit intermolecular lead-sulfur interactions, while allowing the axial coordination of amine Lewis base ligands.
Sigma-Aldrich
2,6-Dimethylbenzenethiol, 95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 kg
1 Available to ship on January 07, 2025
Details...
¥26,700
10 kg
1 Estimated to ship on March 08, 2025
¥212,000