Merck
  • Quantification of the antifungal lipopeptide iturin A by high performance liquid chromatography coupled with aqueous two-phase extraction.
Quantification of the antifungal lipopeptide iturin A by high performance liquid chromatography coupled with aqueous two-phase extraction.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-08-30)
Jun Yuan, Waseem Raza, Qiwei Huang, Qirong Shen
ABSTRACT
Iturin A, a powerful antifungal surfactant, is a kind of bacterial lipopeptide produced by Bacillus strains. This study addresses the use of an aqueous two-phase system (ATPS) using ethanol/ammonium sulfate to extract iturin A from Bacillus amyloliquefaciens NJN-6 fermentation broth and the quantification of iturin A by HPLC. Baseline separation of iturin A homologues was performed using an RP-C(18) column with a mixture of water and acetonitrile. The results showed that the correlation coefficient between integral area and concentration was 0.9961 within the range of 20-140 mg/l. The RSD of the retention time and the peak area were 1.29% and 1.45%, respectively. The effects of some operating parameters in ATPS, e.g., pH, temperature and centrifugation time, were also studied. This method can be successfully used for the rapid quantification of iturin A.
Sigma-Aldrich
Iturin A from Bacillus subtilis, ≥95% (HPLC)
Pricing and availability is not currently available.