Merck
  • Cascade synthesis of polyoxygenated 6H,11H-[2]benzopyrano-[4,3-c][1]benzopyran-11-ones.
Cascade synthesis of polyoxygenated 6H,11H-[2]benzopyrano-[4,3-c][1]benzopyran-11-ones.
The Journal of organic chemistry (2007-03-29)
Mikael I Naumov, Sergey A Sutirin, Andrey S Shavyrin, Olga G Ganina, Irina P Beletskaya, Véronique Bourgarel-Rey, Sébastien Combes, Jean-Pierre Finet, Alexey Yu Fedorov
ABSTRACT
2-(methoxymethoxymethyl)aryllead triacetates, obtained in situ from the corresponding arylboronic acids, reacted with 4-hydroxycoumarins, leading to 3-(2-methoxymethoxymethyl)aryl-4-hydroxycoumarin derivatives in good to high yields. These compounds underwent a cascade sequence of reactions, deprotection-halogenation-annulation, to afford polyoxygenated tetracyclic 6H,11H-[2]benzopyrano-[4,3-c] [1]benzopyran-11-ones in good yields. Some compounds showed a moderate cytotoxicity against human epithelial mammary HBL100 cells.
Sigma-Aldrich
4-Hydroxycoumarin, 98%