Merck
  • EXAFS study of Sn(IV) immobilization by hardened cement paste and calcium silicate hydrates.
EXAFS study of Sn(IV) immobilization by hardened cement paste and calcium silicate hydrates.
Environmental science & technology (2003-06-06)
Isabelle Bonhoure, Erich Wieland, André M Scheidegger, Michael Ochs, Dominik Kunz
ABSTRACT
In this study, the immobilization mechanisms of Sn(IV) onto calcium silicate hydrates (C-S-H) and hardened cement paste (HCP) have been investigated by combining wet chemistry experiments with X-ray absorption spectroscopy (XAS). Evidence is presented which demonstrates the formation of a Sn(IV) inner-sphere surface complex on C-S-H with a CaO/SiO2 weight ratio of 0.7. Two possible structural models, implying a corner sharing between the Sn octahedra and Q1 or Q2b Si tetrahedra, have been developed based on the experimentally determined structural parameters. In HCP, the formation of a different type of Sn(IV) inner-sphere complex has been observed, indicating that C-S-H may not be the uptake-controlling phase for Sn(IV) in the cement matrix. An alternative structural model for Sn(IV) binding in HCP has been developed, assuming that ettringite is the uptake-controlling phase. At high Sn(IV) concentrations, Sn(IV) immobilization in HCP occurs due to the formation of CaSn(OH)6.
Sigma-Aldrich
Tin(IV) chloride, 99.995% trace metals basis
Sigma-Aldrich
Tin(IV) chloride, 98%
Sigma-Aldrich
Tin(IV) chloride solution, 1.0 M in methylene chloride