Merck
  • Targeting de novo lipogenesis and the Lands cycle induces ferroptosis in KRAS-mutant lung cancer.
Targeting de novo lipogenesis and the Lands cycle induces ferroptosis in KRAS-mutant lung cancer.
Nature communications (2022-07-27)
Caterina Bartolacci, Cristina Andreani, Gonçalo Vale, Stefano Berto, Margherita Melegari, Anna Colleen Crouch, Dodge L Baluya, George Kemble, Kurt Hodges, Jacqueline Starrett, Katerina Politi, Sandra L Starnes, Daniele Lorenzini, Maria Gabriela Raso, Luisa M Solis Soto, Carmen Behrens, Humam Kadara, Boning Gao, Ignacio I Wistuba, John D Minna, Jeffrey G McDonald, Pier Paolo Scaglioni
ABSTRACT
Mutant KRAS (KM), the most common oncogene in lung cancer (LC), regulates fatty acid (FA) metabolism. However, the role of FA in LC tumorigenesis is still not sufficiently characterized. Here, we show that KMLC has a specific lipid profile, with high triacylglycerides and phosphatidylcholines (PC). We demonstrate that FASN, the rate-limiting enzyme in FA synthesis, while being dispensable in EGFR-mutant or wild-type KRAS LC, is required for the viability of KMLC cells. Integrating lipidomic, transcriptomic and functional analyses, we demonstrate that FASN provides saturated and monounsaturated FA to the Lands cycle, the process remodeling oxidized phospholipids, such as PC. Accordingly, blocking either FASN or the Lands cycle in KMLC, promotes ferroptosis, a reactive oxygen species (ROS)- and iron-dependent cell death, characterized by the intracellular accumulation of oxidation-prone PC. Our work indicates that KM dictates a dependency on newly synthesized FA to escape ferroptosis, establishing a targetable vulnerability in KMLC.
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Anti-c-K-Ras Antibody, clone 234-4.2, clone 234-4.2, from mouse
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized