Merck
  • A Promising Biomarker and Therapeutic Target in Patients with Advanced PDAC: The Stromal Protein βig-h3.
A Promising Biomarker and Therapeutic Target in Patients with Advanced PDAC: The Stromal Protein βig-h3.
Journal of personalized medicine (2022-04-24)
Christelle de la Fouchardière, Pia Gamradt, Sylvie Chabaud, Maxime Raddaz, Ellen Blanc, Olivier Msika, Isabelle Treilleux, Sophie Bachy, Anne Cattey-Javouhey, Pierre Guibert, Matthieu Sarabi, Pauline Rochefort, Pamela Funk-Debleds, Clélia Coutzac, Isabelle Ray-Coquard, Patrice Peyrat, Pierre Meeus, Michel Rivoire, Aurélien Dupré, Ana Hennino
ABSTRACT
With an overall survival rate of 2-9% at 5 years, pancreatic ductal adenocarcinoma (PDAC) is currently the fourth leading cause of cancer-related deaths in the industrialized world and is predicted to become the second by 2030. Owing to often late diagnosis and rare actionable molecular alterations, PDAC has not yet benefited from the recent therapeutic advances that immune checkpoint inhibitors (ICI) have provided in other cancer types, except in specific subgroups of patients presenting with tumors with high mutational burden (TMB) or microsatellite instability (MSI). The tumor microenvironment (TME) plays a substantial role in therapeutic resistance by facilitating immune evasion. An extracellular stromal protein, βig-h3/TGFβi, is involved in the pathogenesis of PDAC by hampering T cell activation and promoting stiffness of the TME. The study BIGHPANC included 41 patients with metastatic PDAC, and analyzed βig-h3 levels in serum and tumor samples to assess the βig-h3 prognostic value. βig-h3 serum levels are significantly associated with overall survival (HR 2.05, 95%CI 1.07-3.93; p = 0.0301). Our results suggest that βig-h3 serum levels may be considered a prognostic biomarker in patients with metastatic PDAC.
Sigma-Aldrich
Anti-TGFBI antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution