Merck
  • Suppression of the hyaluronic acid pathway induces M1 macrophages polarization via STAT1 in glioblastoma.
Suppression of the hyaluronic acid pathway induces M1 macrophages polarization via STAT1 in glioblastoma.
Cell death discovery (2022-04-13)
Tao Yan, Kaikai Wang, Jiafeng Li, Hong Hu, He Yang, Meng Cai, Ruijie Liu, Honglei Li, Ning Wang, Ying Shi, Wei Hua, Huailei Liu
ABSTRACT
Immunosuppressive tumor microenvironment is a crucial factor that impedes the success of tumor immunotherapy, and tumor-associated macrophages (TAMs) are essential for the formation of tumor immunosuppressive microenvironment. Hyaluronic acid (HA) is highly important brick for glioblastoma microenvironment, but whether it contributes to TAM polarization and glioblastoma immunosuppressive microenvironment is less well known. In our study, we observed that disrupting glioblastoma HA synthesis or blocking HA binding to its receptor CD44 on macrophages increased the proportion of M1 macrophages by upregulating SIRPα in macrophages, the underlying mechanism was elevated SIRPα enhanced STAT1 phosphorylation and suppressed STAT3 phosphorylation in macrophages. Subsequently, the induced macrophages could inhibit glioblastoma growth via a feedback effect. In addition, 4-methylumbelliferone (4MU), a cholecystitis drug, can disrupt the CD47/SIRPα axis by disturbing glioblastoma HA synthesis. Collectively, these findings indicated that HA plays a crucial role in macrophages polarization and CD47/SIRPα signaling between glioblastoma cells and macrophages, and suppressing the HA pathway may be a new immunotherapeutic approach for glioblastoma.
Sigma-Aldrich
Anti-CD44 Rat mAb (A020), liquid, clone A020, Calbiochem®