Merck
  • Controlled phospholipid functionalization of single-walled carbon nanotubes.
Controlled phospholipid functionalization of single-walled carbon nanotubes.
Biomacromolecules (2005-09-13)
Peng He, Marek W Urban
ABSTRACT
These studies show that single-walled carbon nanotubes (SWNTs) can be effectively modified using phospholipids. Using a simple surface modification of SWNTs, followed by deposition of 1,2-bis(10,12-tricosadiynoyl)-sn-glycero-3-phosphoethanolamine (DCPE) phosphipid, results in stable water-dispersible SWNTs with highly uniform thickness.
Product Description
Avanti
7-dehydrocholesterol, Δ,5,7-cholesterol, powder
Avanti
23:2 Diyne PE [DC(8,9)PE], 1,2-bis(10,12-tricosadiynoyl)-sn-glycero-3-phosphoethanolamine, powder