Merck
  • Clinical response and pharmacokinetics of bendamustine as a component of salvage R-B(O)AD therapy for the treatment of primary central nervous system lymphoma (PCNSL).
Clinical response and pharmacokinetics of bendamustine as a component of salvage R-B(O)AD therapy for the treatment of primary central nervous system lymphoma (PCNSL).
BMC cancer (2018-07-11)
Therasa Kim, He Yun Choi, Hyun-Seo Lee, Sung-Hoon Jung, Jae-Sook Ahn, Hyeoung-Joon Kim, Je-Jung Lee, Hee-Doo Yoo, Deok-Hwan Yang
ABSTRACT
A relatively high proportion of patients diagnosed with primary CNS lymphoma will experience recurrent disease, yet therapy options are limited in salvage therapy. This is the first study to evaluate a bendamustine-based combination regimen for the treatment of relapsed/refractory PCNSL and to characterize bendamustine pharmacokinetics in the human CSF. Patients received bendamustine 75 mg/m2 for two days as part of R-B(O)AD administered intravenously every 4 weeks for up to 4 cycles. Response and adverse events of the regimen were assessed. A sparse sampling strategy and population based modeling approach was utilized for evaluation of plasma and CSF levels of bendamustine. Ten patients were enrolled into study of whom 70% were of refractory disease and with high IELSG prognostic risk scores. The ORR of R-BOAD was 50% (95% CI, 0.24 to 0.76) with one patient achieving CR and four PR. Primary toxicity of the regimen was reversible myelosuppression, mostly grade 3 or 4 neutropenia. The Cmax mean for plasma and CSF were 2669 ng/mL and 0.397 ng/mL, respectively, and patients with response at deep tumor sites displayed higher trends in peak exposure. Pharmacokinetic data was best described by a four-compartment model with first-order elimination of drug from central plasma and CSF compartments. R-BOAD is an effective salvage option for PCNSL, but with significant hematologic toxicity. Bendamustine CSF levels are minimal; however correspond to plasma exposure and response. ClinicalTrials.gov NCT03392714 ; retrospectively registered January 8, 2018.
Sigma-Aldrich
Bendamustine hydrochloride hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.